Refine results
  • Show more
Add filters
Adhesives/Sealants
Product Substrate Cure Type Mix Ratio Rheology Work Time Cure System Durometer Data Sheet Comment
MED-1131 Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium RTV 1 PART Thixotropic / non-slump N/A Oxime 35 Type A Download MED-1131 data sheet Non-acidic leaving group
Product Details
Property Average Result
Cure Type RTV
Mix Ratio 1 PART
Rheology Thixotropic / non-slump
Appearance Translucent
Cure 7 days / RTV
Cure System Oxime
Durometer 35 Type A
Elongation 330 %
Flow Rate 0.4 in (10.16 mm)
Specific Gravity 1.1
Substrate Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium
Tack Free Time 20 minutes
Tear 45 ppi (7.94 kN/m)
Tensile 890 psi (6.14 MPa)
Comment Non-acidic leaving group
View more
MED-1137 Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate RTV 1 PART Thixotropic / non-slump N/A Acetoxy 30 Type A Download MED-1137 data sheet Low-slump, solvent-free translucent silicone
Product Details
Property Average Result
Cure Type RTV
Mix Ratio 1 PART
Rheology Thixotropic / non-slump
Appearance Translucent
Cure 72 hours / RTV
Cure System Acetoxy
Durometer 30 Type A
Elongation 560 %
Extrusion Rate 180 g/minute
Flow Rate 0.2 in (5.08 mm)
Specific Gravity 1.07
Stress at Strain 170 psi / 200 % (1.17 MPa / 200 %)
Substrate Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate
Tack Free Time 9 minutes
Tensile 700 psi (4.83 MPa)
Comment Low-slump, solvent-free translucent silicone
View more
MED-1511 Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate / Stainless steel RTV 1 PART Flowable / Self Leveling N/A Acetoxy 25 Type A Download MED-1511 data sheet Low flow
Product Details
Property Average Result
Cure Type RTV
Mix Ratio 1 PART
Rheology Flowable / Self Leveling
Appearance Translucent
Cure 72 hours / RTV
Cure System Acetoxy
Durometer 25 Type A
Elongation 680 %
Extrusion Rate 100 g/minute
Specific Gravity 1.12
Stress at Strain 130 psi / 200 % (0.9 MPa / 200 %)
Substrate Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium / With Primer: Polycarbonate / Stainless steel
Tack Free Time 12 minutes
Tear 95 ppi (16.75 kN/m)
Tensile 1,440 psi (9.93 MPa)
Comment Low flow
View more
MED-1540 Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium RTV 1 PART Flowable / Self Leveling N/A Acetoxy 23 Type A Download MED-1540 data sheet High flow
Product Details
Property Average Result
Cure Type RTV
Mix Ratio 1 PART
Rheology Flowable / Self Leveling
Appearance Translucent
Cure 72 hours / RTV
Cure System Acetoxy
Durometer 23 Type A
Elongation 340 %
Flow Rate 2.7 in (68.58 mm)
Specific Gravity 1.03
Substrate Primerless: Aluminum / Glass / Polyurethane / Silicone / Titanium
Tack Free Time 9.5 minutes
Tear 17 ppi (3 kN/m)
Tensile 265 psi (1.83 MPa)
Comment High flow
View more
MED-2000 Primerless: Glass / Polyurethane / Aluminum / Silicone / Titanium / With Primer: Polycarbonate RTV 1 PART Flowable / Self Leveling N/A Acetoxy 25 Type A Download MED-2000 data sheet Medium flow, high extrusion rate
Product Details
Property Average Result
Cure Type RTV
Mix Ratio 1 PART
Rheology Flowable / Self Leveling
Appearance Translucent
Cure 72 hours / RTV
Cure System Acetoxy
Durometer 25 Type A
Elongation 800 %
Extrusion Rate 2.5 g/minute
Specific Gravity 1.08
Stress at Strain 105 psi / 200 % (0.72 MPa / 200 %)
Substrate Primerless: Glass / Polyurethane / Aluminum / Silicone / Titanium / With Primer: Polycarbonate
Tack Free Time 13 minutes
Tear 80 ppi (14.11 kN/m)
Tensile 1,375 psi (9.48 MPa)
Comment Medium flow, high extrusion rate
View more
MED1-4213 Primerless: Aluminum / Glass / PMMA / Polyurethane / Silicone / Polycarbonate / With Primer: ABS / Polycarbonate / Polyimide / Polysulphone / Polyurethane / PEEK / Polyetherimide / Stainless steel Heat Cure / RTV 1:1 Thixotropic / non-slump 15 minutes Platinum 20 Type A Download MED1-4213 data sheet Quick cure, high extrusion rate
Product Details
Property Average Result
Cure Type Heat Cure / RTV
Mix Ratio 1:1
Rheology Thixotropic / non-slump
Work Time 15 minutes
Appearance Translucent
Cure 24 hours / 25 °C
Cure System Platinum
Durometer 20 Type A
Elongation 800 %
Specific Gravity 1.12
Substrate Primerless: Aluminum / Glass / PMMA / Polyurethane / Silicone / Polycarbonate / With Primer: ABS / Polycarbonate / Polyimide / Polysulphone / Polyurethane / PEEK / Polyetherimide / Stainless steel
Tear 130 ppi (22.93 kN/m)
Tensile 1,000 psi (6.9 MPa)
Viscosity 80,000 cP (80,000 mPa*s)
Comment Quick cure, high extrusion rate
View more
MED2-4213 Primerless: Aluminum / Glass / PMMA / Silicone / With Primer: Plastic / Polycarbonate / Polyetherimide / Polysulphone / PEEK / Stainless steel / Polyimide / Polyurethane Heat Cure 1:1 Thixotropic / non-slump 15 hours Platinum 15 Type A Download MED2-4213 data sheet Quick cure, high extrusion rate
Product Details
Property Average Result
Cure Type Heat Cure
Mix Ratio 1:1
Rheology Thixotropic / non-slump
Work Time 15 hours
Appearance Translucent
Cure 15 minutes / 150 °C
Cure System Platinum
Durometer 15 Type A
Elongation 800 %
Specific Gravity 1.12
Substrate Primerless: Aluminum / Glass / PMMA / Silicone / With Primer: Plastic / Polycarbonate / Polyetherimide / Polysulphone / PEEK / Stainless steel / Polyimide / Polyurethane
Tear 130 ppi (22.93 kN/m)
Tensile 1,000 psi (6.9 MPa)
Viscosity 80,000 cP (80,000 mPa*s)
Comment Quick cure, high extrusion rate
View more
MED3-4213 Primerless: Aluminum / Glass / PMMA / Polycarbonate / Polyurethane / Silicone / With Primer: ABS / PEEK / Polycarbonate / Polyetherimide / Polyimide / Polysulphone / Polyurethane / Stainless steel Heat Cure / RTV 1:1 Thixotropic / non-slump 60 minutes Platinum 20 Type A Download MED3-4213 data sheet Quick cure, high extrusion rate
Product Details
Property Average Result
Cure Type Heat Cure / RTV
Mix Ratio 1:1
Rheology Thixotropic / non-slump
Work Time 60 minutes
Appearance Translucent
Cure 24 hours / 25 °C
Cure System Platinum
Durometer 20 Type A
Elongation 800 %
Specific Gravity 1.12
Substrate Primerless: Aluminum / Glass / PMMA / Polycarbonate / Polyurethane / Silicone / With Primer: ABS / PEEK / Polycarbonate / Polyetherimide / Polyimide / Polysulphone / Polyurethane / Stainless steel
Tear 130 ppi (22.93 kN/m)
Tensile 1,000 psi (6.9 MPa)
Viscosity 80,000 cP (80,000 mPa*s)
Comment Quick cure, high extrusion rate
View more