Refine results
  • Show more
Add filters
Fluids
Product Substrate Volatile Content Viscosity Data Sheet Comment
MED-361 Glass / Metal / Plastic 0.1 % 100 cP / 350 cP / 1,000 cP / 12,500 cP Download MED-361 data sheet Dimethyl Polymer
Product Details
Property Average Result
Volatile Content 0.1 %
Viscosity 100 cP / 350 cP / 1,000 cP / 12,500 cP (100 mPa*s / 350 mPa*s / 1,000 mPa*s / 12,500 mPa*s)
Appearance Clear
Substrate Glass / Metal / Plastic
Comment Dimethyl Polymer
View more
MED-400 Silicone 0.3 % 350 cP / 1,000 cP / 12,500 cP Download MED-400 data sheet Fluorosilicone Polymer
Product Details
Property Average Result
Volatile Content 0.3 %
Viscosity 350 cP / 1,000 cP / 12,500 cP (350 mPa*s / 1,000 mPa*s / 12,500 mPa*s)
Substrate Silicone
Comment Fluorosilicone Polymer
View more
MED-420 Silicone 0.1 % 350 cP / 1,000 cP / 12,500 cP Download MED-420 data sheet Methyl Fluoro Copolymer (Low Fluoro Content)
Product Details
Property Average Result
Volatile Content 0.1 %
Viscosity 350 cP / 1,000 cP / 12,500 cP (350 mPa*s / 1,000 mPa*s / 12,500 mPa*s)
Substrate Silicone
Comment Methyl Fluoro Copolymer (Low Fluoro Content)
View more
MED-460 Silicone 0.1 % 350 cP / 1,000 cP / 12,500 cP Download MED-460 data sheet Methyl Fluoro Copolymer (High Fluoro Content)
Product Details
Property Average Result
Volatile Content 0.1 %
Viscosity 350 cP / 1,000 cP / 12,500 cP (350 mPa*s / 1,000 mPa*s / 12,500 mPa*s)
Substrate Silicone
Comment Methyl Fluoro Copolymer (High Fluoro Content)
View more
Greases
Product Substrate Volatile Content Viscosity Data Sheet Comment
MED-6731 Silicone 0.6 % 2,000,000 cP Download MED-6731 data sheet High Consistency Methyl Fluoro Grease
Product Details
Property Average Result
Volatile Content 0.6 %
Viscosity 2,000,000 cP (2,000,000 mPa*s)
Substrate Silicone
Comment High Consistency Methyl Fluoro Grease
View more
MED-9011 Glass / Metal / Plastic 0.3 % 95,000 cP Download MED-9011 data sheet Low Consistency Grease
Product Details
Property Average Result
Volatile Content 0.3 %
Viscosity 95,000 cP (95,000 mPa*s)
Appearance Translucent
Non Volatile Content 99.4 %
Specific Gravity 1
Substrate Glass / Metal / Plastic
Comment Low Consistency Grease
View more
MED-9021 Glass / Metal / Plastic 0.5 % 300,000 cP Download MED-9021 data sheet Medium Consistency Grease
Product Details
Property Average Result
Volatile Content 0.5 %
Viscosity 300,000 cP (300,000 mPa*s)
Appearance White
Specific Gravity 1.08
Substrate Glass / Metal / Plastic
Comment Medium Consistency Grease
View more
MED-9031 Glass / Metal / Plastic 0.3 % 900,000 cP Download MED-9031 data sheet High Consistency Grease
Product Details
Property Average Result
Volatile Content 0.3 %
Viscosity 900,000 cP (900,000 mPa*s)
Appearance White
Specific Gravity 1.11
Substrate Glass / Metal / Plastic
Comment High Consistency Grease
View more